Ferrocenemethanol 97%
|
Warning
|
Synonyms:
Hydroxymethylferrocene
AC34803-5000
AC34803-0050
AC34803-0010
103466-628EA
99.4
USD
103466-628
103466-626
200063-722
Ferrocenemethanol 97%
Ferrocenemethanol
Specification Test Results
Appearance | Yellow to golden to orange Powder or needles |
Infrared spectrum | Conforms |
Melting point | 77 to 82 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...