Methyltrioxorhenium(VII) 98%

Synonyms: MTOMethylrhenium Trioxide(VII)

AC34696-0025 AC34696-5000
103465-952EA 111 USD
103465-952 103465-950
Methyltrioxorhenium(VII) 98%
Methyltrioxorhenium(VII)
Formula: CH₃O₃Re
MW: 249.24 g/mol
Melting Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00192332
CAS Number: 70197-13-6

Order Now

Specification Test Results

Appearance White to yellow to green to grey to blue-grey Powder or crystals
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR