Acriflavine hydrochloride

Danger

Synonyms: 3,6-Diamino-10-methylacridinium chloride hydrochlorideEuflavine

AC34684-1000 AC34684-0250 AC34684-0100
200036-462EA 217 USD
200036-462 200017-998 200030-296
Acriflavine hydrochloride
Acriflavine hydrochloride

Appearance: Orange-red to red-brown Powder

Formula: C₂₇H₂₈Cl₄N₆
MW: 578.36 g/mol
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00069039
CAS Number: 69235-50-3
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Orange-red to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤7 % (105 °C, 1 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR