Carminic acid 70-90%
|
Danger
|
AC34663-0010
AC34663-0050
200036-770EA
306
USD
200036-770
200036-768
Carminic acid 70-90%
Carminic acid
Formula:
C₂₂H₂₀O₁₃ Melting Pt: 136 °C (dec.) Storage Temperature: Ambient |
EINECS:
215-023-3 UN: 3147 ADR: 8,III |
Specification Test Results
Appearance | Red Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...