1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt hydrate, pure indicator grade

Warning

Synonyms: Nitroso-R salt

AC34655-0250
103518-726EA 31.8 USD
103518-726
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt hydrate, pure indicator grade
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt
Formula: C₁₀H₅NNa₂O₈S₂·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149246
CAS Number: 525-05-3
EINECS: 208-369-1

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 % (1 g, 105 °C)
Suitability (for Co determination) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR