2-Naphthonitrile 97%

Danger

Synonyms: Naphthalene-2-carbonitrile

AC34631-0100 AC34631-0050
200022-856EA 102 USD
200022-856 103530-114
2-Naphthonitrile 97%
2-Naphthonitrile
Formula: C₁₁H₇N
MW: 153.18 g/mol
Boiling Pt: 156…158 °C (12 mmHg)
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00016807
CAS Number: 613-46-7
EINECS: 210-344-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR