Iron(III) sulfate pentahydrate 97%

Danger

Synonyms: ferric sulphate hexahydrateferric sulfate heptahydrateferric sulfateferric sulfate hexahydrateferric sulphateferric sulphate heptahydrate

AC34523-5000 AC34523-0051 AC34523-1000
103529-126EA 81.1 USD
103529-126 103527-734 103527-736
Iron(III) sulfate pentahydrate 97%
Iron(III) sulfate pentahydrate
Formula: Fe₂H₁₀O₁₇S₃
MW: 489.933 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149716
CAS Number: 142906-29-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Appearance Off-white to yellow to beige to grey Crystalline or granular powder
Degree of hydration May vary
Titration with Na2S2O3 20.7 to 23 % (as Fe)
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤200 ppm
Phosphate (PO4) ≤30 ppm
Arsenic (As) ≤5 ppm
Copper (Cu) ≤20 ppm
Zinc (Zn) ≤50 ppm
Ferrous iron (Fe 2+) ≤200 ppm
Insoluble matter ≤0.01 % (in HCl)
Subst not pptd by NH3 ≤0.05 % (as SO4)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR