N,N-Dimethylglycine 97%

Warning

Synonyms: (Dimethylamino)acetic acid2-(Dimethylamino)acetic acid

AC34520-0250 AC34520-0050
200013-524EA 190 USD
200013-524 200007-102
N,N-Dimethylglycine 97%
N,N-Dimethylglycine
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: 175…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 175 to 181 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR