3-Aminopropylphosphonic acid 97%

Warning

AC34494-0010
200016-942EA 134 USD
200016-942
3-Aminopropylphosphonic acid 97%
3-Aminopropylphosphonic acid
Formula: C₃H₁₀NO₃P
MW: 139.0902 g/mol
Melting Pt: 290…294 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008222
CAS Number: 13138-33-5

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR