Methylboronic acid 97%
|
Warning
|
AC34467-0050
AC34467-0010
102775-910EA
142
USD
102775-910
102774-308
Methylboronic acid 97%
Methylboronic acid
Formula:
CH₅BO₂ Melting Pt: 88…90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002105 CAS Number: 13061-96-6 |
Specification Test Results
Appearance | White to beige Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 87 to 91 °C |
Titration with NaOH | 96.0 to 104.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...