3-Carboxyphenylboronic acid 97%

Warning

Synonyms: 3-Carboxybenzeneboronic acid3-Boronobenzoic acid

AC34463-0010 AC34463-0050
200029-224EA 37.5 USD
200029-224 200009-862
3-Carboxyphenylboronic acid 97%
3-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94 g/mol
Melting Pt: 243…247 °C
Storage Temperature: Ambient
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR