(R)-(+)-Hydroxypyrrolidine 98%
|
Warning
|
Synonyms:
(R)-(+)-3-Pyrrolidinol
, (R)-(+)-3-Hydroxypyrrolidine
, (R)-Pyrrolidin-3-ol
AC34457-0010
AC34457-0050
200003-428EA
195
USD
200003-428
200008-214
(R)-(+)-Hydroxypyrrolidine 98%
(R)-(+)-Hydroxypyrrolidine
Formula:
C₄H₉NO MW: 87.12 g/mol Boiling Pt: 108…110 °C (8 mmHg) Flash Pt: >110 °C Storage Temperature: Freezer |
MDL Number:
MFCD00145220 CAS Number: 2799-21-5 |
Specification Test Results
Appearance | Clear yellow to orange-brown Viscous liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4870 to 1.4910 (20 °C, 589 nm) |
GC | ≥97.5 % |
Specific optical rotation | +4.5° to +7.5° (20 °C, 589 nm) (c=3.5, CH3OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...