N-Boc-L-prolinol 98%

Warning

Synonyms: (S)-1-Boc-2-pyrrolidinemethanol(S)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanolN-Boc-L-ProlinolN-(tert-Butoxycarbonyl)-L-prolinol

AC34456-0050 AC34456-0250
103466-202EA 490 USD
103466-202 200013-760
N-Boc-L-prolinol 98%
N-Boc-L-prolinol
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00066232
CAS Number: 69610-40-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 58 to 63 °C
GC ≥97.5 %
Specific optical rotation -46° to -52° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR