Oxypurinol 99%

Danger

Synonyms: 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine

AC33994-0250 AC33994-0050 AC33994-0010
200023-544EA 60 USD
200023-544 200003-418 103530-098
Oxypurinol 99%
Oxypurinol

Appearance: Light yellow Powder

Formula: C₅H₄N₄O₂
MW: 152.113 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00047205
CAS Number: 2465-59-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR