4-(4-Hydroxyphenyl)benzonitrile 95%

Warning

Synonyms: 4'-Hydroxybiphenyl-4-carbonitrile4'-Hydroxy-[1,1'-biphenyl]-4-carbonitrile4'-Hydroxy-4-biphenylcarbonitrile

AC33834-0010
200035-932EA 36.7 USD
200035-932
4-(4-Hydroxyphenyl)benzonitrile 95%
4-(4-Hydroxyphenyl)benzonitrile
Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 192…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00059625
CAS Number: 19812-93-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 192 to 199 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR