Dimethyl(2-hydroxyethyl)phosphonate 95%

Warning

Synonyms: (2-Hydroxyethyl)phosphonic Acid Dimethyl Ester

AC33816-0050 AC33816-0250
200037-144EA 125 USD
200037-144 200037-142
Dimethyl(2-hydroxyethyl)phosphonate 95%
Dimethyl(2-hydroxyethyl)phosphonate

Appearance: Clear colorless to light yellow Liquid

Formula: C₄H₁₁O₄P
MW: 154.10 g/mol
Boiling Pt: 98…104 °C (.1 mmHg)
Density: 1.247 g/cm³
Flash Pt: 192 °C
Storage Temperature: Ambient
MDL Number: MFCD00043376
CAS Number: 54731-72-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4460 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR