(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%

Danger

Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane(1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine

AC33783-0010 AC33783-0050
200003-406EA 296 USD
200003-406 103529-588
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 95%
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine

Appearance: White to light yellow Low melting solid

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00671527
CAS Number: 68737-65-5
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
GC ≥94 %
Specific optical rotation -137° to -149° (20 °C, 589 nm) (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR