(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone 97%

Warning

Synonyms: (S)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one(S)-(+)-5-Hydroxymethyltetrahydro-2-furanone(S)-(+)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone

AC33764-0010 AC33764-2500
200015-364EA 162 USD
200015-364 103513-758
(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone 97%
(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 110…115 °C (2 mmHg)
Density: 1.23 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066224
CAS Number: 32780-06-6

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4680 to 1.4720 (20 °C, 589 nm)
GC ≥96.0 %
Specific optical rotation +30.5° to +36.5° (20 °C, 589 nm) (c=3, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR