(4-Nitrophenyl)-α-D-glucopyranoside 99%

Synonyms: 4-Nitrophenyl α-D-glucopyranoside

AC33715-0250 AC33715-0010 AC33715-0050
200054-740EA 57 USD
200054-740 103513-746 200036-048
(4-Nitrophenyl)-α-D-glucopyranoside 99%
(4-Nitrophenyl) α-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
Melting Pt: 204…218 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064088
CAS Number: 3767-28-0
EINECS: 223-189-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR