D-(-)-Luciferin

Warning

Synonyms: (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid

AC33705-0100 AC33705-0500
103513-744EA 230 USD
103513-744 200018-966
D-(-)-Luciferin
D(-)-Luciferin
Formula: C₁₁H₈N₂O₃S₂
MW: 280.3228 g/mol
Melting Pt: 200 °C
Storage Temperature: Freezer
MDL Number: MFCD00042929
CAS Number: 2591-17-5
EINECS: 219-981-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR