4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate ≥99%

Synonyms: 4-MUP trihydrateDisodium 4-methylumbelliferyl phosphate trihydrate

AC33704-2500
200042-128EA 44.7 USD
200042-128
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate ≥99%
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
Formula: C₁₀H₇Na₂O₆P·3H₂O
MW: 354.16 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037614
CAS Number: 22919-26-2
EINECS: 245-325-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR