4-Methylumbelliferyl-β-D-glucopyranoside 99%

Synonyms: 4-Methylumbelliferyl beta-D-GlucopyranosideMUGlucosid4-MU-β-D-Glc

AC33702-0025 AC33702-5000
103513-742EA 670 USD
103513-742 200017-640
4-Methylumbelliferyl-β-D-glucopyranoside 99%
4-Methylumbelliferyl-beta-D-glucopyranoside
Formula: C₁₆H₁₈O₈
MW: 338.312 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Freezer
MDL Number: MFCD00063694
CAS Number: 18997-57-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
TLC ≥98.5 %
Appearance of solution (3% soln. in acetone/water (2:1)) Clear, colorless
Specific optical rotation -97° to -101° (20 °C, 589 nm) (c=0.3, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR