Dowex® 50WX8 (H) 200-400 mesh ion-exchange resin

Danger

Synonyms: Dowex 50 wx8

AC33535-5000 AC33535-1000 AC33535-0025
200012-288EA 279 USD
200012-288 200006-412 103467-348
Dowex® 50WX8 (H) 200-400 mesh ion-exchange resin
Dowex® 50WX8 (H)
Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 11119-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR