Ethyl-1-hydroxycyclopropanecarboxylate 90%

Synonyms: Ethyl 1-hydroxycyclopropanecarboxylate

AC33530-0010
200003-396EA 182 USD
200003-396
Ethyl-1-hydroxycyclopropanecarboxylate 90%
Ethyl-1-hydroxycyclopropanecarboxylate
Formula: C₆H₁₀O₃
MW: 130.143 g/mol
Boiling Pt: 78 °C (15 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD01863257
CAS Number: 137682-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR