(S)-(+)-Ethyl-3-hydroxybutyrate ≥99%

Synonyms: Ethyl (S)-(+)-3-hydroxybutyrateEthyl (S)-3-hydroxybutyrate(S)-Ethyl-3-hydroxybutyrate(S)-Ethyl 3-hydroxybutanoate

AC33526-0010
200031-206EA 96 USD
200031-206
(S)-(+)-Ethyl-3-hydroxybutyrate ≥99%
(S)-(+)-Ethyl 3-hydroxybutyrate
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 180…182 °C (760 mmHg)
Density: 1.01
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00066206
CAS Number: 56816-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR