1-Allylimidazole 97%

Danger

Synonyms: 1-Allyl-1H-imidazole

AC33517-1000 AC33517-0250
200021-734EA 95 USD
200021-734 200015-684
1-Allylimidazole 97%
1-Allylimidazole
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 212…213 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00037894
CAS Number: 31410-01-2
EINECS: 250-618-1
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR