Ethyl phenoxyacetate 98%

Synonyms: Ethyl 2-phenoxyacetate

AC33494-0050 AC33494-0250 AC33494-1000
103513-686EA 153.1 USD
103513-686 103513-684 200049-434
Ethyl phenoxyacetate 98%
Ethyl phenoxyacetate
Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 136 °C (19 mmHg)
Density: 1.1 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
EINECS: 219-867-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR