1-(1-Cyclohexen-1-yl)pyrrolidine 95%

Warning

Synonyms: 1-Pyrrolidino-1-cyclohexene

AC33483-1000 AC33483-0250
200027-510EA 158 USD
200027-510 200021-510
1-(1-Cyclohexen-1-yl)pyrrolidine 95%
1-(1-Cyclohexen-1-yl)pyrrolidine
Formula: C₁₀H₁₇N
Boiling Pt: 114…115 °C (15 mmHg)
Flash Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003163
CAS Number: 1125-99-1
EINECS: 214-414-6
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR