D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol 96%

Warning

Synonyms: (1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol(1R,2R)-(-)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

AC33479-0250 AC33479-0050
200015-230EA 116.56 USD
200015-230 200045-236
D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol 96%
D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol

Appearance: Light yellow Powder

Formula: C₉H₁₂N₂O₄
MW: 212.21 g/mol
Melting Pt: 160…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00078126
CAS Number: 716-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR