(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 98%

Warning

Synonyms: (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol(S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol(S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline

AC33473-0010
200047-590EA 100.6 USD
200047-590
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 98%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol

Appearance: Off-white Crystalline powder and/or chunks

Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD01631316
CAS Number: 18881-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR