2-Naphthalenethiol 98+%

Warning

Synonyms: 2-Thionaphthol

AC33452-0050 AC33452-0250
200016-710EA 51.4 USD
200016-710 200022-850
2-Naphthalenethiol 98+%
2-Naphthalenethiol
Formula: C₁₀H₈S
MW: 160.24 g/mol
Boiling Pt: 286 °C
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00004086
CAS Number: 91-60-1
EINECS: 202-082-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR