Piperonyl butoxide 90% tech.

Warning

Synonyms: 5-((2-(2-Butoxyethoxy)ethoxy)methyl)-6-propylbenzo[d][1,3]dioxole5-[2-(2-Butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxoleNSC 8401

AC33416-1000 AC33416-5000
200026-486EA 51.1 USD
200026-486 200038-532
Piperonyl butoxide 90% tech.
Piperonyl butoxide
Formula: C₁₉H₃₀O₅
MW: 338.44 g/mol
Boiling Pt: 180 °C (1 mmHg)
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00005842
CAS Number: 51-03-6
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR