Methyl indole-4-carboxylate 99%

Warning

AC33403-0010
200037-936EA 47.7 USD
200037-936
Methyl indole-4-carboxylate 99%
Methyl indole-4-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191222
CAS Number: 39830-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR