Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Supplier: Thermo Scientific
AC33391-5000
200032-280EA 289 USD
200032-280
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Proteins and Peptides
  • Infrared spectrum: conforms
  • Specific optical rotation: –14 to –18° (20 °C, 589 nm) (C=1, 1 N HCl)
  • GHS symbol and signal word: GHS07 and danger
  • EINECS: 261-529-2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR