Carboxymethyl cellulose sodium salt, M.W. ∼ 250000 (DS=1.2)

Synonyms: Carboxymethyl cellulose, sodium saltCellulose, carboxymethyl ether, sodium saltCMCCellulose gumSodium carboxymethyl cellulose

AC33263-0010 AC33263-1000
200030-662EA 103 USD
200030-662 200024-688
Carboxymethyl cellulose sodium salt, M.W. ∼ 250000 (DS=1.2)
Carboxymethyl cellulose sodium salt
Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR