(S)-(-)-2-Methoxypropionic acid 98%

Danger

Synonyms: (S)-2-Methoxypropanoic acid

AC33172-0025 AC33172-5000
200008-208EA 493 USD
200008-208 200008-238
(S)-(-)-2-Methoxypropionic acid 98%
(S)-(-)-2-Methoxypropionic acid

Appearance: Clear colorless to light yellow Liquid

Formula: C₄H₈O₃
MW: 104.105 g/mol
Boiling Pt: 99 °C (20 mmHg)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD01632589
CAS Number: 23953-00-6
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR