1-(6-Methoxy-2-naphthyl)ethanol 98%

Synonyms: 1-(6-Methoxynaphthalen-2-yl)ethanolDL-6-Methoxy-α-methyl-2-naphthalenemethanol

AC33150-0250 AC33150-0050
200025-174EA 69.4 USD
200025-174 200049-212
1-(6-Methoxy-2-naphthyl)ethanol 98%
1-(6-Methoxy-2-naphthyl)ethanol
Formula: C₁₃H₁₄O₂
MW: 202.25 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD01632581
CAS Number: 77301-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR