Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-5-methyl-o-anisidine ≥85%, tech.

Supplier: ACROS ORGANICS MS

Warning
Synonyms: 4-Chloro-2-methoxy-5-methylaniline

AC33136-0050
200017-442EA 19.09 USD InStock
200017-442
4-Chloro-5-methyl-o-anisidine ≥85%, tech.
4-Chloro-5-methyl-o-anisidine


Formula: C₈H₁₀ClNO
MW: 171.63 g/mol
Storage Temperature: Ambient
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Black Crystalline solid
Infrared spectrum Conforms
Melting point 97°C to 103°C
GC ≥80 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty