2-Chloro-p-benzoquinone 95%

Warning

Synonyms: 2-Chloro-1,4-benzoquinone2-Chlorocyclohexa-2,5-diene-1,4-dione

AC33122-0050
103464-448EA 70.3 USD
103464-448
2-Chloro-p-benzoquinone 95%
2-Chloro-p-benzoquinone
Formula: C₆H₃ClO₂
MW: 142.54 g/mol
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00075260
CAS Number: 695-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR