2-Chloromalondialdehyde 95%

Warning

Synonyms: 2-Chloromalonaldehyde

AC33012-0100 AC33012-0500 AC33012-0010
103513-520EA 1006 USD
103513-520 200022-574 200009-324
2-Chloromalondialdehyde 95%
2-Chloromalondialdehyde
Formula: C₃H₃ClO₂
MW: 106.51 g/mol
Melting Pt: 140…145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320489
CAS Number: 36437-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR