n-Dodecyl-β-D-maltoside 99%, high purity

Synonyms: Dodecyl-β-D-maltopyranoside(2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triolLauryl-β-D-maltosideDDM

AC32937-0250 AC32937-0010
200003-368EA 185 USD
200003-368 103466-126
n-Dodecyl-β-D-maltoside 99%, high purity
n-Dodecyl-β-D-maltoside
Formula: C₂₄H₄₆O₁₁
MW: 510.6 g/mol
Melting Pt: 217 °C
Storage Temperature: Freezer
MDL Number: MFCD00043012
CAS Number: 69227-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR