Diethyl (aminomethyl)phosphonate oxalate 95%

Warning

Synonyms: Diethyl (aminomethyl)phosphonate ethanedioatediethyl aminomethylphosphonate oxalatePhosphonic acid, P-(aminomethyl)-, diethyl ester, ethanedioate (1:1)

AC33221-0050 AC33221-0010
200012-066EA 192 USD
200012-066 103513-540
Diethyl (aminomethyl)phosphonate oxalate 95%
Diethyl (aminomethyl)phosphonate oxalate
Formula: C₇H₁₆NO₇P
MW: 257.179 g/mol
Melting Pt: 118…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00800255
CAS Number: 117196-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR