K-252a 98%

Synonyms: Methyl-6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo[b,h]cycloocta[1,2,3,4-jkl]cyclopenta[e]-as-indacene-6-carboxylate

AC32831-1000
200037-720EA 531 USD
200037-720
K-252a 98%
K-252a

Appearance: White to light yellow Powder

Formula: C₂₇H₂₁N₃O₅
MW: 467.481 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00161522
CAS Number: 97161-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR