Diisopropyl azodicarboxylate 94%

Warning

Synonyms: Diisopropyl diazene-1,2-dicarboxylate

AC32756-1000 AC32756-0050 AC32756-5000 AC32756-0250
103527-660EA 420 USD
103527-660 200006-306 200003-342 200003-340
Diisopropyl azodicarboxylate 94%
Diisopropyl azodicarboxylate
Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
Boiling Pt: 75 °C (.2 mmHg)
Melting Pt: 3…5 °C
Density: 1.02 g/cm³
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008875
CAS Number: 2446-83-5
EINECS: 219-502-8
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR