Methyl-4-amino-3-chlorobenzoate 97%

Warning

Synonyms: Methyl 4-amino-3-chlorobenzoate

AC32639-0050 AC32639-0010
200055-866EA 152 USD
200055-866 103512-362
Methyl-4-amino-3-chlorobenzoate 97%
Methyl 4-amino-3-chlorobenzoate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01320686
CAS Number: 84228-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR