Ethyl-3,5-bis(trifluoromethyl)benzoate 97%

Warning

AC32636-0050
200049-342EA 89.1 USD
200049-342
Ethyl-3,5-bis(trifluoromethyl)benzoate 97%
Ethyl 3,5-bis(trifluoromethyl)benzoate
Formula: C₁₁H₈F₆O₂
MW: 286.17
Storage Temperature: Ambient
MDL Number: MFCD01320684
CAS Number: 96617-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR