1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt 95%

Synonyms: 1,3,6,8-Pyrenetetrasulfonic acid tetrasodium salt1,3,6,8-Pyrenetetrasulphonic acid tetrasodium salt

AC32556-0050 AC32556-0010
200027-614EA 80.1 USD
200027-614 200027-616
1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt 95%
1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt

Appearance: Green Crystalline powder

Formula: C₁₆H₆Na₄O₁₂S₄
MW: 610.411 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263184
CAS Number: 59572-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR