2-(N-Ethylanilino)ethanol 96%

Warning

Synonyms: Ethylphenylethanolamine2-(Ethyl(phenyl)amino)ethanolN-Ethyl-N-2-ethanolanilineN-Ethyl-N-(2-hydroxyethyl)aniline

AC32522-1000
200038-184EA 111 USD
200038-184
2-(N-Ethylanilino)ethanol 96%
2-(N-Ethylanilino)ethanol
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Boiling Pt: 276 °C
Melting Pt: 33 °C
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00020575
CAS Number: 92-50-2
EINECS: 202-160-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR