(Trimethylstannyl)diphenylmethylsilane 96%

Danger

AC32521-0010
200039-062EA 46.4 USD
200039-062
(Trimethylstannyl)diphenylmethylsilane 96%
(Trimethylstannyl)diphenylmethylsilane
Formula: C₁₆H₂₂SiSn
Boiling Pt: 125…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00800634
CAS Number: 149013-84-3
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR