Chloro(diisopropyl)phosphine 96%

Danger

Synonyms: Di-i-propylchlorophosphine

AC32155-0250 AC32155-0050
200024-730EA 73.6 USD
200024-730 103527-616
Chloro(diisopropyl)phosphine 96%
Chloro(diisopropyl)phosphine
Formula: C₆H₁₄ClP
MW: 152.6 g/mol
Boiling Pt: 69 °C (33 mmHg)
Density: 0.95 g/cm³
Flash Pt: 3 °C
Storage Temperature: Ambient
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR